Assay Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029522.2 | NP_083798.2 | 12 - 13 | 1947 | 82 | |
| GenBank mRNA | AK018084.1 | - | 2 - 3 | 194 | 82 | |
| AK040996.1 | - | 12 - 13 | 1836 | 82 | ||
| AK143552.1 | - | 12 - 13 | 1948 | 82 | ||
| AY081187.1 | - | 12 - 13 | 1791 | 82 | ||
| BC021308.1 | - | 11 - 12 | 1502 | 5007832 | 82 | |
More Information
Set Membership: |
|

Gpsm2
Genomic Map