Assay Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134114.2 | NP_598875.2 | 1 - 2 | 185 | 68 | |
| GenBank mRNA | AK151338.1 | - | 1 - 2 | 116 | 68 | |
| BC006777.1 | - | 1 - 2 | 140 | 68 | ||
| BC091770.1 | - | 1 - 2 | 186 | 4974098 | 68 | |
| BC130232.1 | - | 1 - 2 | 109 | 40141936 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sft2d1
Genomic Map