Assay Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134114.2 | NP_598875.2 | 2 - 3 | 274 | 62 | |
| GenBank mRNA | AK151338.1 | - | 2 - 3 | 205 | 62 | |
| BC006777.1 | - | 2 - 3 | 229 | 62 | ||
| BC091770.1 | - | 2 - 3 | 275 | 4974098 | 62 | |
| BC130232.1 | - | 2 - 3 | 198 | 40141936 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sft2d1
Genomic Map