Assay Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134114.2 | NP_598875.2 | 7 - 8 | 566 | 83 | |
| GenBank mRNA | AK017471.1 | - | 5 - 6 | 310 | 83 | |
| AK087027.1 | - | 2 - 3 | 109 | 83 | ||
| AK151338.1 | - | 7 - 8 | 497 | 83 | ||
| AK171776.1 | - | 5 - 6 | 331 | 83 | ||
| BC006777.1 | - | 6 - 7 | 1956 | 83 | ||
| BC091770.1 | - | 7 - 8 | 567 | 4974098 | 83 | |
| BC130232.1 | - | 7 - 8 | 490 | 40141936 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974671 | AK151338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974671 | AK171776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974671 | BC006777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974671 | BC091770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974671 | BC130232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139886 | NR_015372.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139886 | BB837600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139886 | AK151338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139886 | BC006777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139886 | NM_134114.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687088 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170385 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12937 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974671 | AK151338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974671 | AK171776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974671 | BC006777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974671 | BC091770.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974671 | BC130232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139886 | NR_015372.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139886 | BB837600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139886 | AK151338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139886 | BC006777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139886 | NM_134114.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03822476 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09554063 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09554048 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153611 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09554022 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03822458 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09554018 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03822457 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09554044 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03822479 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09554038 | - | - |
| Whole Mouse Genome G4122A | A_51_P271074 | BB837600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271074 | BC006777.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109414 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123305 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153611 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32957363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958665 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10453570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441649 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441656 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453564 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441647 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441648 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460656_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385382 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10453562 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10441646 | - | - |
| Mouse Genome 430 2.0 Array | 1460656_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1460656_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700000720.mm.1 | NM_134114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000720.mm.1 | NM_134114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000720.mm.1 | TC1700002764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700000721.mm.1 | NM_134114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000721.mm.1 | NM_134114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000721.mm.1 | TC1700002764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700000724.mm.1 | NM_134114.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000724.mm.1 | NM_134114.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000724.mm.1 | TC1700002764.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460656_a_at | - | - |
| Mouse Expression Set 430 | 1460656_PM_a_at | - | - |
| Murine Genome U74v2 Set | 103863_at | - | - |
| Murine Genome U74v2 Set | 103863_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460656_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460656_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 607662 | - | - |
| ABI Mouse Genome Survey Microarray | 924153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5080402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5080402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773241 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773241 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229765 | - | - |
| mouse-6 v1.1 (Illumina) | scl0106489.6_320-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080402 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00106489.1_143-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0106489.6_320-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360040 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080402 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773241 | - | - |

Sft2d1
Genomic Map