Assay Details
| Gene Symbol: Hmgxb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134134.2 | NP_598895.2 | 2 - 3 | 610 | 104 | |
| NM_178277.1 | NP_840061.1 | 2 - 3 | 609 | 104 | ||
| GenBank mRNA | AB045322.1 | - | 2 - 3 | 610 | 104 | |
| AK034086.1 | - | 2 - 3 | 588 | 104 | ||
| AK041866.1 | - | 2 - 3 | 588 | 104 | ||
| AK144993.1 | - | 2 - 3 | 594 | 104 | ||
| BC079580.1 | - | 2 - 3 | 557 | 5702202 | 104 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB045322.1 transcripts.
Set Membership: |
|

Hmgxb3
Genomic Map