Assay Details
| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144532.2 | NP_653115.1 | 5 - 6 | 840 | 78 | |
| GenBank mRNA | AK010631.1 | - | 2 - 3 | 382 | 78 | |
| AK171863.1 | - | 8 - 9 | 2782 | 78 | ||
| AY039218.1 | - | 5 - 6 | 835 | 78 | ||
| BC029002.1 | - | 4 - 5 | 2080 | 78 | ||
| BC049263.1 | - | 5 - 6 | 840 | 5366999 | 78 | |
| BC053330.1 | - | 4 - 5 | 2127 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187798 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731570 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921303 | - | - |
| Whole Mouse Genome G4122A | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | BC049263.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | BC053330.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | BC029002.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | AK171863.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P625321 | AY039218.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122435 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118106 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14385196 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14385186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14385197 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14385191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14385195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14314045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14239837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14385188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14385196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14250732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14375344 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10464615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10464613 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456936_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425878_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10464612 | - | - |
| Mouse Genome 430 2.0 Array | 1456936_at | - | - |
| Mouse Genome 430 2.0 Array | 1456936_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425878_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425878_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73660_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73660_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900007836.mm.1 | NM_144532.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007836.mm.1 | NM_144532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007836.mm.1 | TC1900000904.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425878_at | - | - |
| Mouse Expression Set 430 | 1425878_PM_at | - | - |
| Mouse Expression Set 430 | 1456936_PM_at | - | - |
| Mouse Expression Set 430 | 1456936_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425878_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425878_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425878_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425878_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456936_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456936_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1660471 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660471 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2713354 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2713354 | - | - |
| mouse-6 v1.1 (Illumina) | scl073660.4_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660471 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl073660.4_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660471 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2713354 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2713353 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2713353 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2713354 | - | - |

Cabp4
Genomic Map