Assay Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144801.2 | NP_659050.2 | 2 - 3 | 267 | 89 | |
| GenBank mRNA | AK035836.1 | - | 2 - 3 | 255 | 89 | |
| AK039173.1 | - | 1 - 2 | 337 | 89 | ||
| AK087636.1 | - | 1 - 2 | 472 | 89 | ||
| AK161481.1 | - | 1 - 2 | 251 | 89 | ||
| BB666121.1 | - | 2 - 3 | 267 | 89 | ||
| BC017618.1 | - | 2 - 3 | 261 | 4923610 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem143
Genomic Map