Assay Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144801.2 | NP_659050.2 | 4 - 5 | 571 | 84 | |
| GenBank mRNA | AK039173.1 | - | 3 - 4 | 641 | 84 | |
| AK087636.1 | - | 3 - 4 | 776 | 84 | ||
| BC017618.1 | - | 4 - 5 | 565 | 4923610 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem143
Genomic Map