| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700007704.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007704.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007704.mm.1 |
TC0700000868.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007707.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007707.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007707.mm.1 |
TC0700000868.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007701.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007701.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007701.mm.1 |
TC0700000868.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007703.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007703.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007703.mm.1 |
TC0700000868.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007708.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007708.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007708.mm.1 |
TC0700000868.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007709.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007709.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007709.mm.1 |
TC0700000868.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007705.mm.1 |
NM_144801.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007705.mm.1 |
NM_144801.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007705.mm.1 |
TC0700000868.mm.1 |
MTA TC |