Assay Details
| Gene Symbol: Aarsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144829.1 | NP_659078.1 | 6 - 7 | 707 | 93 | |
| GenBank mRNA | AK136332.1 | - | 6 - 7 | 674 | 93 | |
| AK168281.1 | - | 6 - 7 | 674 | 93 | ||
| BC005711.1 | - | 6 - 7 | 707 | 3497962 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aarsd1
Genomic Map