Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 12 - 13 | 972 | 101 | |
| GenBank mRNA | AK146202.1 | - | 12 - 13 | 930 | 101 | |
| AK162251.1 | - | 12 - 13 | 921 | 101 | ||
| BC019163.1 | - | 12 - 13 | 952 | 3711115 | 101 | |
| BC021510.1 | - | 12 - 13 | 953 | 5347277 | 101 | |
| BC023680.1 | - | 12 - 13 | 954 | 5343341 | 101 | |
| BC023897.1 | - | 12 - 13 | 929 | 5321187 | 101 | |
| BC027124.1 | - | 12 - 13 | 916 | 5355631 | 101 | |
More Information
Set Membership: |
|

Nae1
Genomic Map