Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 13 - 14 | 1110 | 94 | |
| GenBank mRNA | AK146202.1 | - | 13 - 14 | 1068 | 94 | |
| BC019163.1 | - | 13 - 14 | 1090 | 3711115 | 94 | |
| BC021510.1 | - | 13 - 14 | 1091 | 5347277 | 94 | |
| BC023680.1 | - | 13 - 14 | 1092 | 5343341 | 94 | |
| BC023897.1 | - | 13 - 14 | 1067 | 5321187 | 94 | |
| BC027124.1 | - | 13 - 14 | 1054 | 5355631 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nae1
Genomic Map