Assay Details
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146226.2 | NP_666338.2 | 8 - 9 | 872 | 62 | |
| GenBank mRNA | AK149713.1 | - | 8 - 9 | 874 | 62 | |
| AK212779.1 | - | 3 - 4 | 290 | 62 | ||
| BC005716.1 | - | 7 - 8 | 998 | 62 | ||
| BC025494.1 | - | 8 - 9 | 854 | 5310043 | 62 | |
| BC026594.1 | - | 3 - 4 | 405 | 62 | ||
More Information
Set Membership: |
|

Apeh
Genomic Map