Assay Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290802.1 | NP_001277731.1 | 2 - 3 | 232 | 76 | |
| NM_007645.4 | NP_031671.1 | 1 - 2 | 181 | 76 | ||
| GenBank mRNA | AK154250.1 | - | 1 - 2 | 218 | 76 | |
| AK154466.1 | - | 2 - 3 | 202 | 76 | ||
| AK155663.1 | - | 1 - 2 | 158 | 76 | ||
| AK155966.1 | - | 1 - 2 | 146 | 76 | ||
| BC004610.1 | - | 1 - 2 | 138 | 76 | ||
| BC019402.1 | - | 1 - 2 | 218 | 4013070 | 76 | |
| BC031724.1 | - | 1 - 2 | 138 | 76 | ||
| CT010217.1 | - | 1 - 2 | 70 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd37
Genomic Map