Assay Details
| Gene Symbol: Cd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290802.1 | NP_001277731.1 | 7 - 8 | 842 | 96 | |
| NM_001290804.1 | NP_001277733.1 | 6 - 7 | 767 | 96 | ||
| NM_007645.4 | NP_031671.1 | 6 - 7 | 791 | 96 | ||
| GenBank mRNA | AK154250.1 | - | 6 - 7 | 831 | 96 | |
| AK154466.1 | - | 7 - 8 | 812 | 96 | ||
| AK155966.1 | - | 6 - 7 | 756 | 96 | ||
| BC019402.1 | - | 6 - 7 | 4013070 | 96 | ||
| CT010217.1 | - | 6 - 7 | 680 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd37
Genomic Map