Assay Details
| Gene Symbol: Chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007693.1 | NP_031719.1 | 5 - 6 | 573 | 61 | |
| GenBank mRNA | AK144504.1 | - | 5 - 6 | 567 | 61 | |
| BC026554.1 | - | 5 - 6 | 557 | 4990941 | 61 | |
| M64278.1 | - | 5 - 6 | 573 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144504.1 transcripts.
Set Membership: |
|

Chga
Genomic Map