Assay Details
| Gene Symbol: Cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007811.2 | NP_031837.2 | 3 - 4 | 806 | 66 | |
| GenBank mRNA | AF115769.1 | - | 3 - 4 | 730 | 66 | |
| BC012673.1 | - | 3 - 4 | 766 | 4210893 | 66 | |
| CB574526.1 | - | 2 - 3 | 380 | 66 | ||
| Y12657.1 | - | 3 - 4 | 760 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp26a1
Genomic Map