Assay Details
| Gene Symbol: Dhcr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007856.2 | NP_031882.1 | 8 - 9 | 1459 | 57 | |
| GenBank mRNA | AF057368.1 | - | 7 - 8 | 991 | 57 | |
| AK146811.1 | - | 7 - 8 | 982 | 57 | ||
| BC006854.1 | - | 6 - 7 | 973 | 3157987 | 57 | |
| CT010277.1 | - | 6 - 7 | 952 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
lipid metabolic process
steroid biosynthetic process
cholesterol biosynthetic process
steroid metabolic process
cholesterol metabolic process
post-embryonic development
sterol biosynthetic process
cell differentiation
lung development
multicellular organism growth
regulation of cell proliferation
regulation of cholesterol biosynthetic process
oxidation-reduction process
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors, NAD or NADP as acceptor
7-dehydrocholesterol reductase activity
blood vessel development
lipid metabolic process
steroid biosynthetic process
cholesterol biosynthetic process
steroid metabolic process
cholesterol metabolic process
post-embryonic development
sterol biosynthetic process
cell differentiation
lung development
multicellular organism growth
regulation of cell proliferation
regulation of cholesterol biosynthetic process
oxidation-reduction process
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors, NAD or NADP as acceptor
7-dehydrocholesterol reductase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603237 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603238 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603236 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603235 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603239 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022337 | AK146811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022337 | BC006854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022337 | NM_007856.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022337 | AF057368.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227292 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770654 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180310 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147979 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137656 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108393 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022337 | AK146811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022337 | BC006854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022337 | NM_007856.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022337 | AF057368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290986 | AK146811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290986 | NM_007856.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P290986 | AF057368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P535946 | AK146811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P535946 | BC006854.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P535946 | NM_007856.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P535946 | AF057368.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25603225 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01659928 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147979 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108393 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137656 | - | - |
| Whole Mouse Genome G4122A | A_52_P535946 | AK146811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P535946 | BC006854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P535946 | NM_007856.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P535946 | AF057368.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290986 | AK146811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290986 | NM_007856.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290986 | AF057368.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138183 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108393 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147979 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06584248 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603239 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603235 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603237 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603236 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25603238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25603272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06240947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06591568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06240956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06240950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06591569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06253319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25603216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06473284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06083159 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559315 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559316 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559314 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559313 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559321 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448619_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559312 | - | - |
| Mouse Genome 430 2.0 Array | 1442696_at | - | - |
| Mouse Genome 430 2.0 Array | 1448619_at | - | - |
| Mouse Genome 430 2.0 Array | 1448619_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442696_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13360_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13360_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700018965.mm.1 | NM_007856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018965.mm.1 | NM_007856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018965.mm.1 | TC0700002102.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018964.mm.1 | NM_007856.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018964.mm.1 | NM_007856.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018964.mm.1 | TC0700002102.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448619_PM_at | - | - |
| Mouse Expression Set 430 | 1448619_at | - | - |
| Murine Genome U74v2 Set | 98989_PM_at | - | - |
| Murine Genome U74v2 Set | 98989_at | - | - |
| Murine Genome U74v2 Set | 171506_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171506_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448619_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448619_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448619_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448619_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 851403 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2230148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660471 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2660471 | - | - |
| mouse-6 v1.1 (Illumina) | scl31848.11_248-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31848.11_248-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2660471 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2660471 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31848.11_248-S | - | - |

Dhcr7
Genomic Map