Assay Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007884.2 | NP_031910.1 | 3 - 4 | 285 | 83 | |
| GenBank mRNA | AK005684.1 | - | 3 - 4 | 286 | 83 | |
| AK015223.1 | - | 3 - 4 | 285 | 83 | ||
| AK137179.1 | - | 2 - 3 | 210 | 83 | ||
| AK157900.1 | - | 536 | 83 | |||
| AK157901.1 | - | 2 - 3 | 547 | 83 | ||
| BC012695.1 | - | 2 - 3 | 257 | 4037028 | 83 | |
| D78274.1 | - | 2 - 3 | 200 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epyc
Genomic Map