Assay Details
| Gene Symbol: Ephx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271402.1 | NP_001258331.1 | 7 - 8 | 992 | 81 | |
| NM_001271403.1 | NP_001258332.1 | 6 - 7 | 837 | 81 | ||
| NM_001271421.1 | NP_001258350.1 | 6 - 7 | 887 | 81 | ||
| NM_007940.4 | NP_031966.2 | 7 - 8 | 912 | 81 | ||
| GenBank mRNA | AK002415.1 | - | 7 - 8 | 962 | 81 | |
| AK142714.1 | - | 7 - 8 | 885 | 81 | ||
| AY098585.1 | - | 6 - 7 | 851 | 81 | ||
| BC015087.1 | - | 7 - 8 | 888 | 4218917 | 81 | |
| EF597241.1 | - | 6 - 7 | 887 | 81 | ||
| L05781.1 | - | 7 - 8 | 825 | 81 | ||
| Z37107.1 | - | 7 - 8 | 842 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephx2
Genomic Map