Assay Details
| Gene Symbol: Epx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007946.2 | NP_031972.2 | 5 - 6 | 720 | 90 | |
| GenBank mRNA | BC052881.1 | - | 5 - 6 | 702 | 90 | |
| D78353.1 | - | 5 - 6 | 726 | 90 | ||
| L77979.1 | - | 5 - 6 | 704 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epx
Genomic Map