Assay Details
| Gene Symbol: Ereg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007950.2 | NP_031976.1 | 3 - 4 | 399 | 70 | |
| GenBank mRNA | BB612312.1 | - | 3 - 4 | 394 | 70 | |
| BC027838.1 | - | 3 - 4 | 375 | 5325124 | 70 | |
| D30782.1 | - | 3 - 4 | 394 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ereg
Genomic Map