Assay Details
| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008391.4 | NP_032417.3 | 3 - 4 | 386 | 105 | |
| GenBank mRNA | AK036652.1 | - | 3 - 4 | 354 | 105 | |
| AK087926.1 | - | 3 - 4 | 372 | 105 | ||
| AK132099.1 | - | 3 - 4 | 399 | 105 | ||
| AK155025.1 | - | 3 - 4 | 371 | 105 | ||
| AK158642.1 | - | 1 - 2 | 102 | 105 | ||
| BC006577.1 | - | 3 - 4 | 379 | 3586586 | 105 | |
| BC110666.1 | - | 3 - 4 | 358 | 4918447 | 105 | |
| J03168.1 | - | 3 - 4 | 341 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf2
Genomic Map