Assay Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008409.2 | NP_032435.2 | 4 - 5 | 695 | 105 | |
| GenBank mRNA | AK014024.1 | - | 4 - 5 | 463 | 105 | |
| AK051956.1 | - | 4 - 5 | 674 | 105 | ||
| AK135229.1 | - | 4 - 5 | 647 | 105 | ||
| AK167351.1 | - | 4 - 5 | 696 | 105 | ||
| BC031420.1 | - | 4 - 5 | 662 | 3668557 | 105 | |
| L38971.1 | - | 4 - 5 | 698 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itm2a
Genomic Map