Assay Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008929.3 | NP_032955.2 | 2 - 3 | 352 | 101 | |
| GenBank mRNA | AK148270.1 | - | 2 - 3 | 309 | 101 | |
| BB618718.1 | - | 2 - 3 | 352 | 101 | ||
| U28423.1 | - | 2 - 3 | 295 | 101 | ||
More Information
Set Membership: |
|

Dnajc3
Genomic Map