Assay Details
| Gene Symbol: Glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009752.2 | NP_033882.1 | 5 - 6 | 673 | 70 | |
| NR_108101.1 | 4 - 5 | 503 | 70 | |||
| NR_108102.1 | 4 - 5 | 503 | 70 | |||
| GenBank mRNA | AK034049.1 | - | 5 - 6 | 652 | 70 | |
| AK034966.1 | - | 474 | 70 | |||
| AK042069.1 | - | 5 - 6 | 643 | 70 | ||
| AK048460.1 | - | 5 - 6 | 604 | 70 | ||
| AK076517.1 | - | 5 - 6 | 644 | 70 | ||
| AK153218.1 | - | 5 - 6 | 656 | 70 | ||
| AK166473.1 | - | 5 - 6 | 658 | 70 | ||
| AK167271.1 | - | 3 - 4 | 283 | 70 | ||
| AK171593.1 | - | 5 - 6 | 676 | 70 | ||
| AK171760.1 | - | 4 - 5 | 474 | 70 | ||
| BC003998.1 | - | 3 - 4 | 244 | 70 | ||
| BC028875.1 | - | 5 - 6 | 593 | 4240285 | 70 | |
| M57734.1 | - | 5 - 6 | 632 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glb1
Genomic Map