Assay Details
| Gene Symbol: Glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009752.2 | NP_033882.1 | 8 - 9 | 1029 | 93 | |
| NR_108101.1 | 7 - 8 | 859 | 93 | |||
| NR_108102.1 | 7 - 8 | 859 | 93 | |||
| GenBank mRNA | AK034049.1 | - | 8 - 9 | 1008 | 93 | |
| AK034966.1 | - | 830 | 93 | |||
| AK042069.1 | - | 8 - 9 | 999 | 93 | ||
| AK048460.1 | - | 8 - 9 | 960 | 93 | ||
| AK076517.1 | - | 8 - 9 | 1000 | 93 | ||
| AK153218.1 | - | 8 - 9 | 1011 | 93 | ||
| AK171593.1 | - | 8 - 9 | 1032 | 93 | ||
| AK171760.1 | - | 7 - 8 | 830 | 93 | ||
| BC028875.1 | - | 8 - 9 | 949 | 4240285 | 93 | |
| M57734.1 | - | 8 - 9 | 988 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glb1
Genomic Map