Assay Details
| Gene Symbol: Glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009752.2 | NP_033882.1 | 11 - 12 | 1266 | 79 | |
| NR_108101.1 | 10 - 11 | 1187 | 79 | |||
| NR_108102.1 | 10 - 11 | 1096 | 79 | |||
| GenBank mRNA | AK034966.1 | - | 1158 | 79 | ||
| AK042069.1 | - | 11 - 12 | 1236 | 79 | ||
| AK076517.1 | - | 11 - 12 | 1237 | 79 | ||
| AK153218.1 | - | 11 - 12 | 1248 | 79 | ||
| AK171593.1 | - | 11 - 12 | 1269 | 79 | ||
| AK171760.1 | - | 10 - 11 | 1067 | 79 | ||
| AK204903.1 | - | 3 - 4 | 244 | 79 | ||
| BC028875.1 | - | 11 - 12 | 1186 | 4240285 | 79 | |
| M57734.1 | - | 11 - 12 | 1225 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glb1
Genomic Map