Assay Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009759.4 | NP_033889.2 | 15 - 16 | 1695 | 65 | |
| GenBank mRNA | AF012104.1 | - | 14 - 15 | 1548 | 65 | |
| AK036707.1 | - | 14 - 15 | 1554 | 65 | ||
| AK040936.1 | - | 15 - 16 | 1696 | 65 | ||
| BC138104.1 | - | 15 - 16 | 1572 | 8861123 | 65 | |
| BC138105.1 | - | 15 - 16 | 1572 | 8861124 | 65 | |
| U88091.1 | - | 15 - 16 | 1681 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmx
Genomic Map