Assay Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009881.3 | NP_034011.1 | 4 - 5 | 436 | 99 | |
| GenBank mRNA | AF081260.1 | - | 4 - 5 | 418 | 99 | |
| AF081261.1 | - | 4 - 5 | 418 | 99 | ||
| BC055103.1 | - | 4 - 5 | 385 | 6741468 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdyl
Genomic Map