Assay Details
| Gene Symbol: Ctsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009983.2 | NP_034113.1 | 4 - 5 | 530 | 68 | |
| GenBank mRNA | AK089294.1 | - | 4 - 5 | 488 | 68 | |
| AK150363.1 | - | 4 - 5 | 491 | 68 | ||
| AK150364.1 | - | 4 - 5 | 488 | 68 | ||
| AK150579.1 | - | 4 - 5 | 488 | 68 | ||
| AK150622.1 | - | 4 - 5 | 488 | 68 | ||
| AK150636.1 | - | 4 - 5 | 488 | 68 | ||
| AK150694.1 | - | 4 - 5 | 488 | 68 | ||
| AK150768.1 | - | 4 - 5 | 488 | 68 | ||
| AK150946.1 | - | 4 - 5 | 540 | 68 | ||
| AK151256.1 | - | 4 - 5 | 488 | 68 | ||
| AK151275.1 | - | 4 - 5 | 488 | 68 | ||
| AK151276.1 | - | 4 - 5 | 488 | 68 | ||
| AK151374.1 | - | 4 - 5 | 488 | 68 | ||
| AK151829.1 | - | 4 - 5 | 488 | 68 | ||
| AK151962.1 | - | 4 - 5 | 491 | 68 | ||
| AK152036.1 | - | 4 - 5 | 501 | 68 | ||
| AK152045.1 | - | 4 - 5 | 541 | 68 | ||
| AK152168.1 | - | 4 - 5 | 488 | 68 | ||
| AK152187.1 | - | 4 - 5 | 488 | 68 | ||
| AK152188.1 | - | 4 - 5 | 488 | 68 | ||
| AK152406.1 | - | 4 - 5 | 491 | 68 | ||
| AK152458.1 | - | 4 - 5 | 488 | 68 | ||
| AK152567.1 | - | 4 - 5 | 488 | 68 | ||
| AK152578.1 | - | 4 - 5 | 488 | 68 | ||
| AK152593.1 | - | 4 - 5 | 518 | 68 | ||
| AK152680.1 | - | 4 - 5 | 488 | 68 | ||
| AK152828.1 | - | 4 - 5 | 488 | 68 | ||
| AK152881.1 | - | 4 - 5 | 488 | 68 | ||
| AK152892.1 | - | 4 - 5 | 488 | 68 | ||
| AK153191.1 | - | 4 - 5 | 488 | 68 | ||
| AK153291.1 | - | 4 - 5 | 488 | 68 | ||
| AK159097.1 | - | 4 - 5 | 488 | 68 | ||
| AK159209.1 | - | 4 - 5 | 488 | 68 | ||
| AK159440.1 | - | 4 - 5 | 541 | 68 | ||
| AK159490.1 | - | 4 - 5 | 488 | 68 | ||
| AK159512.1 | - | 4 - 5 | 530 | 68 | ||
| AK159580.1 | - | 4 - 5 | 488 | 68 | ||
| AK159642.1 | - | 2 - 3 | 240 | 68 | ||
| AK159743.1 | - | 4 - 5 | 492 | 68 | ||
| AK159896.1 | - | 4 - 5 | 488 | 68 | ||
| AK159970.1 | - | 4 - 5 | 541 | 68 | ||
| AK165227.1 | - | 4 - 5 | 558 | 68 | ||
| AK166864.1 | - | 4 - 5 | 501 | 68 | ||
| AK166938.1 | - | 4 - 5 | 535 | 68 | ||
| AK167143.1 | - | 4 - 5 | 526 | 68 | ||
| AK167366.1 | - | 4 - 5 | 560 | 68 | ||
| AK167698.1 | - | 4 - 5 | 487 | 68 | ||
| AK167867.1 | - | 4 - 5 | 489 | 68 | ||
| AK169139.1 | - | 4 - 5 | 541 | 68 | ||
| AK171195.1 | - | 4 - 5 | 489 | 68 | ||
| AK172583.1 | - | 4 - 5 | 488 | 68 | ||
| AK194256.1 | - | 1 - 2 | 58 | 68 | ||
| AK213259.1 | - | 1 - 2 | 58 | 68 | ||
| AK213518.1 | - | 1 - 2 | 58 | 68 | ||
| BC054758.1 | - | 4 - 5 | 560 | 6413315 | 68 | |
| BC057931.1 | - | 4 - 5 | 521 | 5253319 | 68 | |
| X52886.1 | - | 4 - 5 | 530 | 68 | ||
| X53337.1 | - | 4 - 5 | 470 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsd
Genomic Map