Assay Details
| Gene Symbol: Ackr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF016584.1 | - | 1 - 2 | 60 | 86 | |
| AF109159.1 | - | 1 - 2 | 119 | 86 | ||
| BC005583.1 | - | 1 - 2 | 118 | 3968984 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF016584.1, AF109159.1, BC005583.1 transcripts.
Set Membership: |
|

Ackr1
Genomic Map