Assay Details
| Gene Symbol: Nmur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010341.1 | NP_034471.1 | 1 - 2 | 833 | 58 | |
| GenBank mRNA | AF044602.1 | - | 1 - 2 | 833 | 58 | |
| BC120822.1 | - | 1 - 2 | 854 | 40129745 | 58 | |
| BC137775.1 | - | 1 - 2 | 854 | 8860794 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
chloride transport
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
activation of phospholipase C activity
neuropeptide signaling pathway
chemical synaptic transmission
calcium-mediated signaling
inositol phosphate-mediated signaling
positive regulation of protein targeting to mitochondrion
neuromedin U receptor activity
signal transducer activity
G-protein coupled receptor activity
neuropeptide receptor activity
calcium ion transport
chloride transport
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
activation of phospholipase C activity
neuropeptide signaling pathway
chemical synaptic transmission
calcium-mediated signaling
inositol phosphate-mediated signaling
positive regulation of protein targeting to mitochondrion
neuromedin U receptor activity
signal transducer activity
G-protein coupled receptor activity
neuropeptide receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179577 | BC120822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179577 | BC137775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179577 | NM_010341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179577 | AF044602.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235177 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160971 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179577 | BC120822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179577 | BC137775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179577 | NM_010341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179577 | AF044602.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130965 | BC120822.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130965 | BC137775.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130965 | NM_010341.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130965 | AF044602.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20411859 | - | - |
| Whole Mouse Genome G4122A | A_51_P130965 | BC120822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130965 | BC137775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130965 | NM_010341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130965 | AF044602.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122043 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109247 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00955333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00955328 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01075334 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01075347 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00955335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20411847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00174222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00021081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00174224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00291408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00770582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00978361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01028181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00021078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01075333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00978363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00955335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00021082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01075340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00955336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00829678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01075344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00217724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20411826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00919670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10356336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356337 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421667_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10356335 | - | - |
| Mouse Genome 430 2.0 Array | 1421667_at | - | - |
| Mouse Genome 430 2.0 Array | 1421667_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14767_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14767_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100022180.mm.1 | TC0100002743.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421667_at | - | - |
| Mouse Expression Set 430 | 1421667_PM_at | - | - |
| Murine Genome U74v2 Set | 99417_PM_at | - | - |
| Murine Genome U74v2 Set | 99417_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421667_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421667_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421667_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421667_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 408314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1500403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1500403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256834 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256834 | - | - |
| mouse-6 v1.1 (Illumina) | scl16519.2.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl16519.2.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256834 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256834 | - | - |

Nmur1
Genomic Map