Assay Details
| Gene Symbol: Hdac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077696.1 | NP_001071164.1 | 14 - 15 | 2398 | 90 | |
| NM_001284248.1 | NP_001271177.1 | 16 - 17 | 2910 | 90 | ||
| NM_001284249.1 | NP_001271178.1 | 15 - 16 | 2507 | 90 | ||
| NM_001284250.1 | NP_001271179.1 | 14 - 15 | 2395 | 90 | ||
| NM_010412.3 | NP_034542.3 | 14 - 15 | 2395 | 90 | ||
| GenBank mRNA | AF006602.2 | - | 13 - 14 | 2108 | 90 | |
| AF207748.1 | - | 13 - 14 | 2027 | 90 | ||
| AK122325.2 | - | 9 - 10 | 1437 | 90 | ||
| AK139202.1 | - | 15 - 16 | 2481 | 90 | ||
| AK145972.1 | - | 14 - 15 | 2361 | 90 | ||
| AK146483.1 | - | 14 - 15 | 2334 | 90 | ||
| AK155335.1 | - | 14 - 15 | 2369 | 90 | ||
| AK156008.1 | - | 8 - 9 | 3232 | 90 | ||
| AK157159.1 | - | 8 - 9 | 3232 | 90 | ||
| BC023309.1 | - | 5 - 6 | 948 | 90 | ||
| BC049926.1 | - | 14 - 15 | 2329 | 90 | ||
| BC058554.1 | - | 16 - 17 | 2862 | 90 | ||
| BC060609.1 | - | 13 - 14 | 2189 | 5720438 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdac5
Genomic Map