Assay Details
| Gene Symbol: Hdac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077696.1 | NP_001071164.1 | 22 - 23 | 3167 | 60 | |
| NM_001284248.1 | NP_001271177.1 | 24 - 25 | 3679 | 60 | ||
| NM_001284249.1 | NP_001271178.1 | 23 - 24 | 3276 | 60 | ||
| NM_001284250.1 | NP_001271179.1 | 22 - 23 | 3164 | 60 | ||
| NM_010412.3 | NP_034542.3 | 22 - 23 | 3164 | 60 | ||
| GenBank mRNA | AF006602.2 | - | 21 - 22 | 2877 | 60 | |
| AF207748.1 | - | 21 - 22 | 2796 | 60 | ||
| AK122325.2 | - | 17 - 18 | 2206 | 60 | ||
| AK139202.1 | - | 23 - 24 | 3250 | 60 | ||
| AK145972.1 | - | 22 - 23 | 3130 | 60 | ||
| AK146483.1 | - | 22 - 23 | 3103 | 60 | ||
| AK155335.1 | - | 22 - 23 | 3138 | 60 | ||
| AK161956.1 | - | 2 - 3 | 296 | 60 | ||
| BC023309.1 | - | 13 - 14 | 1717 | 60 | ||
| BC049926.1 | - | 22 - 23 | 3098 | 60 | ||
| BC058554.1 | - | 24 - 25 | 3631 | 60 | ||
| BC060609.1 | - | 21 - 22 | 2958 | 5720438 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdac5
Genomic Map