Assay Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010485.3 | NP_034615.2 | 3 - 4 | 509 | 95 | |
| GenBank mRNA | AK034536.1 | - | 3 - 4 | 496 | 95 | |
| AK075818.1 | - | 3 - 4 | 471 | 95 | ||
| AK078333.1 | - | 3 - 4 | 825 | 95 | ||
| AK080365.1 | - | 3 - 4 | 467 | 95 | ||
| AK089088.1 | - | 2 - 3 | 216 | 95 | ||
| AK148528.1 | - | 3 - 4 | 95 | |||
| AK161667.1 | - | 2 - 3 | 226 | 95 | ||
| AK180399.1 | - | 2 - 3 | 250 | 96 | ||
| AK190746.1 | - | 2 - 3 | 250 | 95 | ||
| AK197032.1 | - | 2 - 3 | 250 | 95 | ||
| AK204358.1 | - | 2 - 3 | 196 | 95 | ||
| AK214281.1 | - | 1 - 2 | 53 | 95 | ||
| U65735.1 | - | 2 - 3 | 278 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elavl1
Genomic Map