Assay Details
| Gene Symbol: Ccl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011332.3 | NP_035462.2 | 1 - 2 | 120 | 76 | |
| GenBank mRNA | AF125570.1 | - | 1 - 2 | 209 | 76 | |
| AF125571.1 | - | 1 - 2 | 151 | 76 | ||
| AF125572.1 | - | 1 - 2 | 124 | 76 | ||
| AJ242587.1 | - | 1 - 2 | 99 | 76 | ||
| AK007663.1 | - | 1 - 2 | 121 | 76 | ||
| BC028505.1 | - | 1 - 2 | 104 | 3328278 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccl17
Genomic Map