Assay Details
| Gene Symbol: Adgre5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163029.1 | NP_001156501.1 | 5 - 6 | 683 | 97 | |
| NM_001163030.1 | NP_001156502.1 | 6 - 7 | 830 | 97 | ||
| NM_001163031.1 | NP_001156503.1 | 5 - 6 | 677 | 97 | ||
| NM_011925.2 | NP_036055.2 | 7 - 8 | 965 | 97 | ||
| GenBank mRNA | AF146344.1 | - | 5 - 6 | 554 | 97 | |
| AK004577.1 | - | 5 - 6 | 693 | 97 | ||
| AK218664.1 | - | 3 - 4 | 357 | 97 | ||
| BC006676.1 | - | 5 - 6 | 545 | 3499451 | 97 | |
| CT010326.1 | - | 5 - 6 | 509 | 97 | ||
| Y18365.1 | - | 7 - 8 | 823 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre5
Genomic Map