Assay Details
| Gene Symbol: Adgre5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163029.1 | NP_001156501.1 | 6 - 7 | 722 | 83 | |
| NM_001163030.1 | NP_001156502.1 | 7 - 8 | 869 | 83 | ||
| NM_001163031.1 | NP_001156503.1 | 6 - 7 | 716 | 83 | ||
| NM_011925.2 | NP_036055.2 | 8 - 9 | 1004 | 83 | ||
| GenBank mRNA | AF146344.1 | - | 6 - 7 | 593 | 83 | |
| AK004577.1 | - | 6 - 7 | 732 | 83 | ||
| BC006676.1 | - | 6 - 7 | 584 | 3499451 | 83 | |
| CT010326.1 | - | 6 - 7 | 548 | 83 | ||
| Y18365.1 | - | 8 - 9 | 862 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre5
Genomic Map