Assay Details
| Gene Symbol: Adgre5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163029.1 | NP_001156501.1 | 9 - 10 | 1079 | 66 | |
| NM_001163030.1 | NP_001156502.1 | 10 - 11 | 1226 | 66 | ||
| NM_001163031.1 | NP_001156503.1 | 9 - 10 | 1073 | 66 | ||
| NM_011925.2 | NP_036055.2 | 11 - 12 | 1361 | 66 | ||
| GenBank mRNA | AF146344.1 | - | 9 - 10 | 950 | 66 | |
| AK004577.1 | - | 9 - 10 | 1089 | 66 | ||
| BC006676.1 | - | 9 - 10 | 941 | 3499451 | 66 | |
| CT010326.1 | - | 9 - 10 | 905 | 66 | ||
| Y18365.1 | - | 11 - 12 | 1219 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgre5
Genomic Map