Assay Details
| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011935.3 | NP_036065.1 | 1 - 2 | 231 | 90 | |
| GenBank mRNA | AB221580.1 | - | 1 - 2 | 65 | 90 | |
| AF117254.1 | - | 1 - 2 | 128 | 90 | ||
| BC082324.1 | - | 1 - 2 | 199 | 6850748 | 90 | |
| CJ062694.1 | - | 1 - 2 | 232 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Esrrg
Genomic Map