Assay Details
| Gene Symbol: Ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013562.2 | NP_038590.2 | 5 - 6 | 800 | 75 | |
| GenBank mRNA | AK078888.1 | - | 798 | 75 | ||
| AK168682.1 | - | 5 - 6 | 797 | 75 | ||
| BC037013.1 | - | 5 - 6 | 774 | 75 | ||
| BC043723.1 | - | 5 - 6 | 767 | 5369100 | 75 | |
| X17400.1 | - | 5 - 6 | 777 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd1
Genomic Map