Assay Details
| Gene Symbol: Chaf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013733.3 | NP_038761.1 | 11 - 12 | 1978 | 78 | |
| GenBank mRNA | AJ132771.2 | - | 11 - 12 | 1882 | 78 | |
| AK034839.1 | - | 11 - 12 | 1979 | 78 | ||
| AK161016.1 | - | 806 | 78 | |||
| BC053740.1 | - | 11 - 12 | 1949 | 6504838 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chaf1a
Genomic Map