Assay Details
| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013755.3 | NP_038783.1 | 2 - 3 | 249 | 98 | |
| GenBank mRNA | AA553085.1 | - | 2 - 3 | 248 | 98 | |
| AF114031.1 | - | 2 - 3 | 184 | 98 | ||
| AK159576.1 | - | 2 - 3 | 230 | 98 | ||
| BC029903.1 | - | 2 - 3 | 210 | 4487769 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gyg
Genomic Map