Assay Details
| Gene Symbol: Irf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016851.2 | NP_058547.2 | 1 - 2 | 185 | 85 | |
| GenBank mRNA | AK087631.1 | - | 1 - 2 | 143 | 85 | |
| AK143299.1 | - | 1 - 2 | 136 | 85 | ||
| AK143954.1 | - | 1 - 2 | 170 | 85 | ||
| AK149487.1 | - | 1 - 2 | 148 | 85 | ||
| BC008515.1 | - | 1 - 2 | 135 | 3592582 | 85 | |
| BC071221.1 | - | 1 - 2 | 177 | 85 | ||
| BY100044.1 | - | 1 - 2 | 186 | 85 | ||
| U73029.1 | - | 1 - 2 | 135 | 85 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK087631.1,AK143299.1,AK143954.1,AK149487.1,BC008515.1,BC071221.1,NM_016851.2,U73029.1 transcripts.
Set Membership: |
|

Irf6
Genomic Map