Assay Details
| Gene Symbol: Hnrnpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170981.1 | NP_001164452.1 | 3 - 4 | 442 | 78 | |
| NM_001170982.1 | NP_001164453.1 | 2 - 3 | 416 | 78 | ||
| NM_001170983.1 | NP_001164454.1 | 3 - 4 | 442 | 78 | ||
| NM_001170984.1 | NP_001164455.1 | 3 - 4 | 442 | 78 | ||
| NM_016884.3 | NP_058580.1 | 3 - 4 | 442 | 78 | ||
| GenBank mRNA | AF095257.1 | - | 3 - 4 | 396 | 78 | |
| AK010845.1 | - | 3 - 4 | 421 | 78 | ||
| AK011336.1 | - | 3 - 4 | 414 | 78 | ||
| AK012633.2 | - | 3 - 4 | 404 | 78 | ||
| AK019958.1 | - | 3 - 4 | 1617 | 78 | ||
| AK088245.1 | - | 3 - 4 | 419 | 78 | ||
| AK088678.1 | - | 3 - 4 | 414 | 78 | ||
| AK089061.1 | - | 3 - 4 | 422 | 78 | ||
| AK151446.1 | - | 3 - 4 | 419 | 78 | ||
| AK153053.1 | - | 3 - 4 | 419 | 78 | ||
| AK166590.1 | - | 3 - 4 | 420 | 78 | ||
| AK168802.1 | - | 3 - 4 | 416 | 78 | ||
| AK169287.1 | - | 3 - 4 | 414 | 78 | ||
| AK184811.1 | - | 3 - 4 | 349 | 78 | ||
| AK212671.1 | - | 1 - 2 | 268 | 78 | ||
| AK214960.1 | - | 2 - 3 | 304 | 78 | ||
| BC004706.1 | - | 3 - 4 | 404 | 3499283 | 78 | |
| BC095922.1 | - | 2 - 3 | 357 | 4988965 | 78 | |
| CK793036.1 | - | 396 | 78 | |||
| DV652725.1 | - | 3 - 4 | 466 | 78 | ||
More Information
Set Membership: |
|

Hnrnpc
Genomic Map