Assay Details
| Gene Symbol: Elovl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019423.2 | NP_062296.1 | 4 - 5 | 377 | 61 | |
| GenBank mRNA | AF170908.1 | - | 4 - 5 | 367 | 61 | |
| AK014803.1 | - | 4 - 5 | 456 | 61 | ||
| AK029855.1 | - | 4 - 5 | 358 | 61 | ||
| AK044770.1 | - | 4 - 5 | 380 | 61 | ||
| AK050397.1 | - | 4 - 5 | 380 | 61 | ||
| BC098215.1 | - | 4 - 5 | 360 | 30617894 | 61 | |
| KF987080.1 | - | 2 - 3 | 275 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elovl2
Genomic Map