Assay Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019517.4 | NP_062390.3 | 6 - 7 | 1044 | 67 | |
| GenBank mRNA | AF051150.1 | - | 2 - 3 | 193 | 67 | |
| AF216310.1 | - | 6 - 7 | 995 | 67 | ||
| AK052309.1 | - | 6 - 7 | 1031 | 67 | ||
| AK078770.1 | - | 6 - 7 | 1011 | 67 | ||
| AK165036.1 | - | 6 - 7 | 980 | 67 | ||
| BC120773.1 | - | 6 - 7 | 1010 | 40129696 | 67 | |
| BC131947.1 | - | 6 - 7 | 1010 | 40130224 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bace2
Genomic Map