Assay Details
| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019537.2 | NP_062410.1 | 6 - 7 | 809 | 99 | |
| GenBank mRNA | AJ238270.1 | - | 6 - 7 | 811 | 99 | |
| AK042203.1 | - | 6 - 7 | 820 | 99 | ||
| AK088118.1 | - | 6 - 7 | 822 | 99 | ||
| BC032965.1 | - | 6 - 7 | 802 | 1196077 | 99 | |
| BC060285.1 | - | 6 - 7 | 823 | 6508084 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990391 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990390 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091671 | AK042203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091671 | AK088118.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091671 | BC032965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091671 | BC060285.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091671 | NM_019537.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134728 | AK042203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134728 | AK088118.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134728 | BC032965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134728 | BC060285.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134728 | NM_019537.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539461 | AK030069.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539461 | AK035313.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539461 | AK220202.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539461 | NM_172854.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477323 | AK082825.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132195 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160892 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117219 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134728 | AK042203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134728 | AK088118.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134728 | BC032965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134728 | BC060285.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134728 | NM_019537.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091671 | AK042203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091671 | AK088118.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091671 | BC032965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091671 | BC060285.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091671 | NM_019537.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30990398 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160892 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117219 | - | - |
| Whole Mouse Genome G4122A | A_51_P421950 | AK042203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421950 | AK088118.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421950 | BC032965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421950 | BC060285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421950 | NM_019537.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134728 | AK042203.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134728 | AK088118.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134728 | BC032965.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134728 | BC060285.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134728 | NM_019537.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118522 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160892 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12873481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12476275 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12873472 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12873474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990391 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30990390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30990396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12627434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12665692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13001047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12805965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12986006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12873489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12873485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30990369 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10441113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441110 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448307_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10441107 | - | - |
| Mouse Genome 430 2.0 Array | 1448307_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448307_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56088_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56088_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600016385.mm.1 | NM_019537.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600016385.mm.1 | TC1600002135.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600016384.mm.1 | NM_019537.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600016384.mm.1 | TC1600002135.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448307_PM_at | - | - |
| Mouse Expression Set 430 | 1448307_at | - | - |
| Murine Genome U74v2 Set | 97220_at | - | - |
| Murine Genome U74v2 Set | 97220_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448307_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448307_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448307_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448307_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1170706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258358 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258358 | - | - |
| mouse-6 v1.1 (Illumina) | scl056088.2_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056088.2_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2874662 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2874662 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056088.2_28-S | - | - |

Psmg1
Genomic Map