Assay Details
| Gene Symbol: Fzr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019757.1 | NP_062731.1 | 3 - 4 | 333 | 72 | |
| GenBank mRNA | AF083809.1 | - | 3 - 4 | 333 | 72 | |
| AK051993.1 | - | 3 - 4 | 415 | 72 | ||
| AK081476.1 | - | 3 - 4 | 387 | 72 | ||
| AK083686.1 | - | 396 | 72 | |||
| AK154822.1 | - | 3 - 4 | 417 | 72 | ||
| AK155370.1 | - | 3 - 4 | 415 | 72 | ||
| AK163951.1 | - | 3 - 4 | 392 | 72 | ||
| AK172679.1 | - | 3 - 4 | 427 | 72 | ||
| BC006616.1 | - | 3 - 4 | 414 | 3967009 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fzr1
Genomic Map